Skip to Content
Merck
All Photos(1)

Key Documents

ATEH95EB77BD

AstaTech, Inc.

2-(1-BENZOYLPIPERIDIN-4-YL)ACETIC ACID

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
247.29
MDL number:

Iupac Name

(1-benzoyl-4-piperidinyl)acetic acid

Inchi Code

1S/C14H17NO3/c16-13(17)10-11-6-8-15(9-7-11)14(18)12-4-2-1-3-5-12/h1-5,11H,6-10H2,(H,16,17)

InChI key

KXHGWYZKPOTYOL-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=204849

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=D84429


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library