Skip to Content
Merck
All Photos(1)

Key Documents

ATEH0DDC4328

AstaTech, Inc.

2-AMINO-4-ISOPROPYLPYRIMIDINE-5-CARBOXYLIC ACID

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
181.19
MDL number:

Iupac Name

2-amino-4-isopropyl-5-pyrimidinecarboxylic acid

Inchi Code

1S/C8H11N3O2/c1-4(2)6-5(7(12)13)3-10-8(9)11-6/h3-4H,1-2H3,(H,12,13)(H2,9,10,11)

InChI key

SIOOCOUQJSQPNO-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=122780

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=F20211


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library