Skip to Content
Merck
All Photos(1)

Key Documents

APO455834327

Apollo Scientific Ltd

2-Chloro-5-(trifluoromethyl)phenacyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
301.49
MDL number:

bp

27-29

Iupac Name

2-bromo-1-[2-chloro-5-(trifluoromethyl)phenyl]ethanone

Inchi Code

1S/C9H5BrClF3O/c10-4-8(15)6-3-5(9(12,13)14)1-2-7(6)11/h1-3H,4H2

InChI key

YVTHTWZIUKRHCF-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/2-chloro-5-trifluoromethylphenacyl-bromide

Synonym

2-Bromo-2'-chloro-5'-(trifluoromethyl)acetophenone, 2-Bromo-1-[2-chloro-5-(trifluoromethyl)phenyl]ethan-1-one

Boiling point

27-29

Pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-chloro-5-trifluoromethylphenacyl-bromide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library