AMBH303C4C1E
Ambeed, Inc.
2,4-Dibromo-1,3,5-trifluorobenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2,4-dibromo-1,3,5-trifluorobenzene
Inchi Code
1S/C6HBr2F3/c7-4-2(9)1-3(10)5(8)6(4)11/h1H
InChI key
SFNQFVCUPVBCOC-UHFFFAOYSA-N
storage temp.
Sealed in dry,2-8C
Purity
97%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
Solid or Semi-solid or liquid
Product Link
https://www.ambeed.com/products/363-69-9.html
Signal Word
Warning
Hazard Statements
Precautionary statements
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.