Documents
TEM-G400F1-CU
Sigma-Aldrich
Gilder Finder Grids G400F1
Copper, size 400 mesh, vial of 100
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
vial of 100
size
400 mesh
General description
Gilder Finder Grids G400F1. Copper grids, 400 mesh. The block of 1152 (32x36) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each small block of 36 (6x6) cells has a central number from 1 to 32 for identification purposes.