Documents
MT30207961
Mettler Toledo Seven2Go Pro conductivity meter S7
Meter Only
About This Item
UNSPSC Code:
41115609
NACRES:
NB.61
description
connector LTW 7-pin (IP67)
interface: micro-USB
pH/ion/ORP meters
feature
Meter Only
manufacturer/tradename
Mettler-Toledo 30207961
TDS measuring range
0.01 mg/L-600 g/L
conductivity measuring range
0.00-2022% (Ash)
0.01 μS/cm-1000 mS/cm
range of application measuring range
0.00-100.0 Ω-cm (specific resistance)
salinity measuring range
0.00-42 (psu - practical salinity units)
temperature measuring range
-5-105 °C
battery power
4 x 1.5 V AA (or 1.3 V NiMH accumulators)
General description
Simple and reliable, attractively designed and robust. The Seven2Go portables are engineered to provide users with fast quality data, one-handed operation, and durability, making them ideally suited for mobile applications in the laboratory, at-line, or outdoors.
Simple Menu – Intuitive Operation:
  • Seven2Go′s new intuitive menu guarantees out-of-the-box operation for anyone. Users no longer need to study lengthy operating instructions
T-Pad – Comfortable, Fast Navigation:
  • Seven2Go′s new T-Pad improves the speed with which you move through menus. The smart button arrangement allows for faster navigation that provide you shorter time to actual measurement
ISM® Ready – Reduced Chances for Error:
  • Upon connection of any InLab ISM (Intelligent Sensor Management) sensor, the instrument automatically recognizes the sensor and stores sensor ID, serial number, and last calibration data
IP67 – Weatherproof and Robust Design:
  • Thanks to its IP67 protection and drop test resistance, the new Seven2Go portables are able to withstand harsh and demanding environments
12 Languages – No more Confusion:
  • The Seven2Go supports 12 languages and offers a user interface that uses complete words and sentences. The lack of cryptic expressions simplify operation and reduces strain on the user
Sunlight-proof – Always Brilliant Readability:
  • The screen′s contrast is enhanced by direct sunlight. This guarantees an excellent readability even under bright and sunny conditions
StatusLight – Easy Readiness Check:
  • Thanks to the new StatusLight you are instantly aware of how the meter is doing. An easy to understand traffic light concept shows if your meter is ready or not
Features and Benefits
Includes: Meter, LabX direct pH, USB cable, electrode clip, base stand, instructions, quick guide, test certificate, declaration of conformity, batteries
Legal Information
ISM is a registered trademark of Mettler-Toledo, Inc.
Seven2Go is a trademark of Mettler-Toledo, Inc.