Skip to Content
Merck
All Photos(1)

Documents

BR402038

BRAND® volumetric flask for sugar analysis

size 100 mL

Synonym(s):

Flasks, SILBERBRAND, Volumetric flasks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

material

glass

feature

class B For Kohlrausch sugar analysis, Calibrated to contain (TC, In)

packaging

pack of 2 ea

manufacturer/tradename

BRAND 402038

size

100 mL

Looking for similar products? Visit Product Comparison Guide

General description

Borosilicate glass 3.3. For Kohlrausch sugar analysis. Error limits according to Class B. Calibrated to contain (TC, In.).

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service