Skip to Content
Merck
All Photos(1)

Key Documents

635820

Sigma-Aldrich

Diethyl cyclopropylmethyl phosphonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5CH2P(O)(OCH2CH3)2
CAS Number:
Molecular Weight:
192.19
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Assay

97%

form

solid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.4380 (lit.)

bp

60 °C/0.125 mmHg (lit.)

density

1.023 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(CC1CC1)OCC

InChI

1S/C8H17O3P/c1-3-10-12(9,11-4-2)7-8-5-6-8/h8H,3-7H2,1-2H3

InChI key

SEZHIFQWGROIOQ-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of alkyldichlorophosphine via chlorination reaction under mild conditions

Structural characteristics:
  • Intramolecular hydrogen binding via "banana" bonds between a H atom belonging to the phosphino group and the edge of the cyclopropyl ring

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service