472042
Glycolic acid ethoxylate octyl ether
average Mn ~410
Synonym(s):
Capryleth-6 carboxylic acid, Capryleth-9 carboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mol wt
average Mn ~410
impurities
≤1.0 wt. % NaCl
refractive index
n20/D 1.45 (lit.)
density
1.04 g/mL at 25 °C
SMILES string
CCCCCCCCOCCOCCOCCOCCOCCOCC(O)=O
Looking for similar products? Visit Product Comparison Guide
Application
Low foaming anionic surfactant.
Features and Benefits
Hydrotope. Stable toward acids, bases and electrolytes.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service