AC7300013066
Advanced Chemical Intermediates Ltd.
Ethyl 2-benzyloxycarbonylamino-3-(toluene-4-sulfonyloxy)propionate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
ethyl 2-{[(benzyloxy)carbonyl]amino}-3-{[(4-methylphenyl)sulfonyl]oxy}propanoate
Inchi Code
1S/C20H23NO7S/c1-3-26-19(22)18(21-20(23)27-13-16-7-5-4-6-8-16)14-28-29(24,25)17-11-9-15(2)10-12-17/h4-12,18H,3,13-14H2,1-2H3,(H,21,23)
InChI key
GFSFXIOOUGXOOH-UHFFFAOYSA-N
storage temp.
Room temperature
Purity
95%
Country of Origin
GB
Shipping Temperature
Room temperature
Physical Form
Beige solid
Product Link
https://www.acints.com/products/ACI-NI-00309
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.acints.com/products/ACI-NI-00309
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.