
Doramectin

Synonym(s):
25-Cyclohexyl-5-O-demethyl-25-de(1-methylpropyl)avermectin
Empirical Formula (Hill Notation):
C50H74O14
CAS Number:
Molecular Weight:
899.11
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing