TEM-Q350AR1.3
QUANTIFOIL R 1.2/1.3
Gold Grids, size 300 mesh, pack of 50
Synonym(s):
R 1.2/1.3 Quantifoil grid
About This Item
Recommended Products
packaging
pack of 50
size
300 mesh
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Pressure-Assisted Cryo-EM Specimen Preparation: This study investigates a pressure-assisted method for preparing cryo-EM specimens using different types of QUANTIFOIL grids, including R 1.2/1.3, to enhance image quality (Lebrette et al., 2019).
- Rapid Single-Particle Cryo-EM: Discusses the use of QUANTIFOIL R 1.2/1.3 grids for rapid data collection in cryo-EM, contributing to advancements in structural biology techniques (Efremov & Stroobants, 2021).
- Structural Analysis of Bovine Thyroglobulin: This research utilizes QUANTIFOIL R 1.2/1.3 grids to compare the structures of human and bovine thyroglobulin, aiding in understanding protein architecture (Kim et al., 2022).
- Optimizing Solution Transfer in Cryo-EM: Explores modifications to QUANTIFOIL grids to enhance solution transfer during cryo-EM experiments, aiming to improve imaging conditions (Chen et al., 2019).
- Laser Diffraction Real-Time Measurement: Focuses on using laser diffraction to measure liquid amounts on QUANTIFOIL grids, presenting a method to control sample application in cryo-EM studies (Ahn et al., 2020).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service