672599
(R)-(+)-1-(2,4,6-Triisopropylphenyl)ethanol
≥96.0% (HPLC)
Synonym(s):
(R)-Stericol®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H28O
CAS Number:
Molecular Weight:
248.40
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥96.0% (HPLC)
optical purity
enantiomeric ratio: ≥98:2 (HPLC)
functional group
hydroxyl
storage temp.
2-8°C
SMILES string
CC(C)c1cc(C(C)C)c([C@@H](C)O)c(c1)C(C)C
InChI
1S/C17H28O/c1-10(2)14-8-15(11(3)4)17(13(7)18)16(9-14)12(5)6/h8-13,18H,1-7H3/t13-/m1/s1
InChI key
VYKPQIIFUMKNLF-CYBMUJFWSA-N
Application
Legal Information
Stericol is a registered trademark of Université Joseph Fourier
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A New Asymmetric Carbon-Carbon Bond Forming Reaction: Four-Component Stereoselective Synthesis of (Z)-4,6-Dihydroxy-3-methylalk-2-enyl Methyl Sulfones This work was supported by the Swiss National Science Foundation.
Narkevitch et al.
Angewandte Chemie (International ed. in English), 39(10), 1806-1808 (2000-08-10)
Caroline Roche et al.
The Journal of organic chemistry, 70(21), 8352-8363 (2005-10-08)
Three natural pyrrolizidines, (+)-amphorogynines A and D and (+)-retronecine, have been prepared from a common lactam intermediate. This central compound, in turn, was synthesized in diastereomerically enriched form through a highly selective [2 + 2]-cycloaddition of dichloroketene with a chiral
Hamel, M., et al.
Tetrahedron Asymmetry, 16, 3406-3415 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service