Merck
HPLC Analysis of Methylselenocysteine Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of Methylselenocysteine Enantiomers on Astec® CHIROBIOTIC® TAG application for HPLC
材料
分析色谱柱
产品编号
说明
价格
Astec® CHIROBIOTIC® TAG 手性 HPLC色谱 柱
5 μm particle size, L × I.D. 25 cm × 4.6 mm
标准
产品编号
说明
价格
Se-(甲基)硒代半胱氨酸 盐酸盐
≥95% (TLC)
CONDITIONS
column
CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 μm particles (14024AST)
mobile phase
[A] water; [B] acetonitrile; (30:70, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV at 230 nm
injection
5 μL
sample
2 mg/mL in methanol