Merck
HPLC Analysis of Dansyl-Norleucine Enantiomers on Astec® CHIROBIOTIC® R
HPLC Analysis of Dansyl-Norleucine Enantiomers on Astec® CHIROBIOTIC® R application for HPLC
材料
分析色谱柱
产品编号
说明
价格
Astec® CHIROBIOTIC® R手性HPLC柱
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 μm particles (13024AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (70:30. A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 260 nm
injection
1 μL
sample
5 mg/mL in methanol