Merck
Trace HPLC Analysis of Proline Enantiomers on Astec® CLC-L
Trace HPLC Analysis of Proline Enantiomers on Astec® CLC-L application for HPLC
材料
分析色谱柱
产品编号
说明
价格
Astec® CLC-L 手性 HPLC 色谱柱
5 μm particle size, L × I.D. 15 cm × 4.6 mm
标准
产品编号
说明
价格
L-脯氨酸
ReagentPlus®, ≥99% (HPLC)
L-脯氨酸
from non-animal source, meets EP, USP testing specifications, suitable for cell culture
DL-脯氨酸
ReagentPlus®, 99%
CONDITIONS
column
Astec CLC-L, 15 cm x 4.6 mm I.D., 5 μm particles (53123AST)
mobile phase
5 mM copper sulfate
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
10 μL
sample
1 mg/mL in water