Merck
HPLC Analysis of Nucleosides on Ascentis® Phenyl
HPLC Analysis of Nucleosides on Ascentis® Phenyl application for HPLC
材料
标准
产品编号
说明
价格
HPLC 核苷类测试混标
in 1% sodium formate (varied conc.), analytical standard
腺苷
Pharmaceutical Secondary Standard; Certified Reference Material
鸟苷酸二钠
Pharmaceutical Secondary Standard; Certified Reference Material
腺苷杂质F
Pharmaceutical Secondary Standard; Certified Reference Material
鸟苷
Pharmaceutical Secondary Standard; Certified Reference Material
腺苷5′-一磷酸
Pharmaceutical Secondary Standard; Certified Reference Material
胸苷
pharmaceutical secondary standard
胞苷
certified reference material, pharmaceutical secondary standard
CONDITIONS
column
Ascentis Phenyl, 15 cm x 2.1 mm I.D., 5 μm particles (581613-U)
mobile phase
10 mM ammonium formate (pH 3.0 with formic acid)
flow rate
0.2 mL/min
column temp.
35 °C
detector
UV, 270 nm; 750 psi back pressure regulator on outlet of flow cell
injection
1 μL
sample
as indicated, in mobile phase