Merck
HPLC Analysis of Proteins on TSKgel® Phenyl-5PW
HPLC Analysis of Proteins on TSKgel® Phenyl-5PW application for HPLC
材料
分析色谱柱
产品编号
说明
价格
TSKgel®疏水作用色谱法(HIC)色谱柱
L × I.D. 7.5 cm × 7.5 mm, 10 μm particle size
CONDITIONS
column
TSKgel Phenyl-5PW, 7.5 cm × 7.5 mm I.D., 10 μm particles (807573)
mobile phase
0.1 M phosphate buffer, pH 7.0 plus ammonium sulfate, 1.8 M to 0 M in 60 min
flow rate
1 mL/min
detector
UV, 280 nm
说明
法律信息
TSKgel is a registered trademark of Tosoh Corporation