Merck
HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® R (MS Detection)
HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® R (MS Detection) application for HPLC
材料
分析色谱柱
产品编号
说明
价格
Astec® CHIROBIOTIC® R手性HPLC柱
5 μm particle size, L × I.D. 15 cm × 2.1 mm
应用
产品编号
说明
价格
LC/MS Analysis of Cannabinoids on Ascentis® RP-Amide
application for HPLC
HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® V
application for HPLC
标准
产品编号
说明
价格
酮洛芬
meets USP testing specifications
酮洛芬
≥98% (TLC)
(S)-(+)-酮洛芬
99%
酮洛芬
VETRANAL®, analytical standard
CONDITIONS
column
CHIROBIOTIC R, 15 cm x 2.1 mm, 5 μm particles (13019AST)
mobile phase
[A] 20 mM ammonium acetate, pH 5.6 [B] methanol; (70:30, A:B)
flow rate
0.2 mL/min
column temp.
35 °C
detector
ESI(-)