Merck
GC Analysis of 1,2,3,4-Tetrahydro-1-Naphthylamine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX B-DA
GC Analysis of 1,2,3,4-Tetrahydro-1-Naphthylamine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX™ B-DA suitable for GC
材料
分析色谱柱
产品编号
说明
价格
Astec® CHIRALDEX B-DA毛细管GC色谱柱
L × I.D. 30 m × 0.25 mm, df 0.12 μm
标准
产品编号
说明
价格
1,2,3,4-四氢萘
analytical standard
1,2,3,4-四氢萘
anhydrous, 99%
CONDITIONS
column
Astec CHIRALDEX B-DA, 30 m x 0.25 mm I.D., 0.12 μm (72023AST)
oven
140 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
sample
peaks 1 & 2: 1,2,3,4-tetrahydro-1-naphthylamine enantiomers (N-TFA derivatives)
说明
法律信息
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC