GC Analysis of FAMEs in Soybean Oil on SLB-IL111
材料
分析柱
产品编号
说明
价格
CONDITIONS
column
SLB-IL111, 30 m x 0.25 mm I.D., 0.20 μm (28927-U)
oven
180 °C
inj. temp.
250 °C
detector
FID, 260 °C
carrier gas
helium, 25 cm/sec
injection
1 μL, 50:1 split
liner
4 mm I.D., split type, cup design
sample
Characterized reference oils, methylated using BF3-methanol
prior to analysis