返回
Ace replacement reactor head, PTFE
Products
制造商
直径(mm)
筛选产品
Showing 1-1 of 1
比较
货号
说明
价格
Z690384
Ace replacement reactor head, PTFE,
4 openings, fits 250 mL and 500 mL PTFE reactors, diam. 60 mm, joint: ST/NS 24/40