Merck
503711
调节器壁支架
Two station, CGA 580
登录查看公司和协议定价
About This Item
UNSPSC代码:
41112500
描述
two pigtails (on brackets with CGA connectors)
特点
Two station
配件
CGA 580
正在寻找类似产品? 访问 产品对比指南
一般描述
Wall-mounting a gas cylinder regulator eliminates the need to handle it during cylinder change-out, thereby reducing the chance of kinking the tubing (kinked tubing next to a fitting may result in a leak). The bracket is fabricated from grade 304 stainless steel for long life, and can be used for 2-stage or 1-stage gas cylinder regulators (order separately). An integral 30 in. flexible stainless steel pigtail allows connection to a cylinder without the risk of a leak caused from kinked tubing. The CGA connector is a hand-tight design, eliminating the need for a wrench during cylinder change-out. Choose:
  • CGA 580 for helium, nitrogen, and argon
  • CGA 350 for hydrogen, ethane, and 5% methane in argon
  • CGA 590 for compressed air
The two station bracket has 2 pigtails that join before the regulator fitting, and allows 2 gas cylinders to feed the system, extending the time between cylinder change-out.
历史批次信息供参考:
分析证书(COA)