2637405
1 of 10
Z420085
STAT-O-LOK™ quick-disconnect
Sigma-Aldrich
C4642
α-环糊精
ACE585306
Ace-Safe® tubing connector, UHDPE
SYNF100002
Synthware™ 47 mm filtration apparatus clamp
Supelco
57258
Visiprep™ 固相萃取装置收集板(除非另外注明,否则已包括在标准和 DL 型号中)
58705
Cheminert® 管道接头
Z420077
Z126551
Automatic shut-off quick-disconnect
58685
无凸缘接头
57161-U
固相萃取装置真空表/真空调节阀
Splitless (unpacked)
pkg of 5 ea
54 mm × 5.0 mm × 2.0 mm
正在寻找类似产品? 访问 产品对比指南
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.