Merck
23783
Needle File
4 in. long, slim tapered files, pkg of 2 ea
登录查看公司和协议定价
About This Item
UNSPSC代码:
41100000
包装
pkg of 2 ea
一般描述
4 in. long, slim taper files.
Use to enlarge ferrule I.D. Also handy for removing ferrules stuck in fittings.
历史批次信息供参考:
分析证书(COA)