主要文件
Z768588
UK cord set for Belly Button®, Belly Dancer® and Low Profile Roller (BS1363 Standard)
pack of 1 ea
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103811
NACRES:
NB.73
包装
pack of 1 ea
制造商/商品名称
IBI COMCD0005
一般描述
This is required to use any of the Belly Button or Belly Dancer products in the UK.
法律信息
The Belly Button is a registered trademark of IBI Scientific Corp.
The Belly Dancer is a registered trademark of IBI Scientific Corp.
历史批次信息供参考:
分析证书(COA)