主要文件
Z719706
Sorenson low binding standard tips
pack of 8 ea (Empty racks), non-sterile
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121600
NACRES:
NB.01
材料
colorless
polypropylene
無菌
non-sterile
包裝
pack of 8 ea (Empty racks)
製造商/商標名
Sorenson 28310
最大體積
200 μL
正在寻找类似产品? 访问 产品对比指南
一般說明
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
法律資訊
Sorenson is a trademark of Sorenson BioScience, Inc.
历史批次信息供参考:
分析证书(COA)