主要文件
OH30073980
Ohaus® Pen pH Meter
Model ST20T-A
登录查看公司和协议定价
About This Item
UNSPSC代码:
41115603
物料
epoxy shaft
包装
pack of 1 ea
制造商/商品名称
Ohaus® 30073980
TDS 测量范围
0.0-100.0 mg/L
温度测量范围
0.0-99.0 °C
电池电量
4 × 1.5 V, 200 hr life
正在寻找类似产品? 访问 产品对比指南
一般描述
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
应用
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
特点和优势
Easily replaceable electrodes, automatic shut off feature
法律信息
Ohaus is a registered trademark of Ohaus Corp.
历史批次信息供参考:
分析证书(COA)