描述
alarm
timing measuring range
100 hr, resolution: 1/100 sec
长度 × 高度 × 直径
61 × 71 × 25 mm
电池电量
1 × 1.5 V (LR44)
一般描述
Pocket, four channel stopwatch / timer with both audible and visual alarms.
特点和优势
- Four independent timing channels with visual and audible alarms
- use as a stopwatch, a count up/down, or a clock
- includes spring clip and magnetic back
法律信息
Clearly Safe is a registered trademark of Heathrow Scientific LLC