Merck
EP6731011055
Eppendorf® CellXpert® C170i CO2 Incubator
Oxygen control, inner door with 4 door segments, handle left side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
登录查看公司和协议定价
About This Item
UNSPSC代码:
41104405
包装
pack of 1 ea
制造商/商品名称
Eppendorf® 6731011055
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
CellXpert® C170i, inner door with 4 door segments, handle left side, 1 – 20 % oxygen control, 100 – 127 V/50 – 60 Hz (US/JP/South America)
特点和优势
  • 1 – 20 % oxygen control
  • handle left side
  • inner door with 4 door segments
  • 180° HTD (standard with C170i)
  • including water tray and four shelves (stainless steel)
  • 100 – 127 V, 50 – 60 Hz
法律信息
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)