Merck
EP5804709004
Eppendorf® Swing bucket rotor A-4-44
incl. 4 x 100 ml rectangular buckets
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103909
NACRES:
NB.82
特點
to hold 4 x 100 mL rectangular buckets
incl. 4 x 100 ml rectangular buckets
製造商/商標名
Eppendorf® 5804709004
參數
5,000 max. RCF (4,500 xg)
技術
centrifugation: suitable
相容性
for use with centrifuge models 5804/5804R/5810/5810R
法律資訊
Eppendorf is a registered trademark of Eppendorf AG