主要文件
EP022822047
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-45-70-11, non aerosol tight, to hold, 70 x 1.5/2.0 mL reaction vessels
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103909
特点
to hold 70 x 1.5/2.0 mL reaction vessels
fixed angle rotor
non aerosol tight
包装
pack of 1 ea
制造商/商品名称
Eppendorf® 022822047 (NA)
技术
centrifugation: suitable
相容性
for use with centrifuge models Vacufuge Plus Concentrator
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Selection of rotors for the Eppendorf Vacufuge Plus.
法律信息
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)