Merck
BR31571
BRAND® BLAUBRAND®量筒,有序号
capacity 10 mL
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
包裝
pkg of 12 ea
製造商/商標名
BRAND 27452
高度
115 mm
容量
10 mL
正在寻找类似产品? 访问 产品对比指南
一般說明
Measuring cylinders tall form. Class A DE-M marking to DIN EN ISO 4788. Made from DURAN(R), calibrated to contain (TC, In). With spout and hexagonal glass base. Ring marks at major graduations. High contrast blue enamel graduations and inscription
法律資訊
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG