主要文件
RX003100
Sigma-Aldrich
Water Bath Pump
for use with Midas Water Bath
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103715
AC/DC 輸入
115 V AC
應用
hematology
microbiology
相容性
for use with Midas Water Bath
一般說明
The Water Bath Pump is a centrifugal pump and is powered by 115V AC. The Water Bath Pump is a spare part for the Midas® Water Bath (64000WB-94). The Midas® Water Bath is an accessory for the Midas® III-Plus Automated Stainer (64000H-94).
法律資訊
MIDAS is a registered trademark of Merck KGaA, Darmstadt, Germany