主要文件
EZFITVANG3
Millipore
EZ-Fit® Vanguard for EZ-FitHTA manifold head
spare part, pkg of 3 units
登录查看公司和协议定价
About This Item
UNSPSC代码:
41116129
NACRES:
NB.24
物料
plastic
质量水平
灭菌适用性
autoclavable compatible
包装
pkg of 3 units
制造商/商品名称
EZ-Fit®
适用性
suitable (bioburden testing)
应用
pharmaceutical (Water testing, Beverage Industry)
相容性
for use with EZ-Fit® Filtration Unit
一般描述
The EZ-Fit® Manifold Head High Throughput Application has been designed to improve ease of use, reduce maintenance and limit the risk of contamination. These filtration heads offer full compatibility with the EZ-Fit® Filtration Units.
法律信息
EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany