主要文件
Z741835
Extech Stem Thermometer, Waterproof
登录查看公司和协议定价
About This Item
分類程式碼代碼:
42182209
NACRES:
NB.79
特點
CE compliant
battery included (LR44)
resolution 0.1 °F/C
(WEEE Compliant)
包裝
pkg of 1 ea
製造商/商標名
Extech Instruments 39240
溫度測量範圍
-40-392 °F (-40 to 200 °C)
尺寸
5.9 in. × 0.8 in. × 0.7 in., 150 x 20 x 18mm
重量
0.7 oz20 g
特點和優勢
  • Pocket size with Max/Min recording
  • Measures temperature in frozen solids and liquids
  • 2.75" (70mm) Stainless steel stem
历史批次信息供参考:
分析证书(COA)