主要文件
Z615633
Julabo® bath carrier trays for SW22/SW23
to fit 8 Erlenmeyer flasks 500 cc
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122800
NACRES:
NB.77
制造商/商品名称
Julabo 8970365
Julabo
正在寻找类似产品? 访问 产品对比指南
一般描述
Erlenmeyer flasks can be positioned in the shaking water bath using the stainless steel tray. A maximum of 8 Erlenmeyer flasks with 500 ml capacity fit on a standard tray.
法律信息
Julabo is a registered trademark of Julabo USA, Inc.
历史批次信息供参考:
分析证书(COA)