主要文件
Z531170
Sigma-Aldrich
Aldrich® coiled condenser
unjacketed, L 400 mm, joint: ST/NS 29/32
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
接合处
joint: ST/NS29/32
长度
400 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)