主要文件
Z243299
Sigma-Aldrich
ACE Squibb separatory funnel with Locktight stopper
capacity 1,000 mL, stopper joint: ST/NS 24/40
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121809
NACRES:
NB.43
接合处
joint: ST/NS stopper24/40
制造商/商品名称
Ace Glass 723163
容量
1,000 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
European style funnel with new leaktight locking stopper that will not fall out during shaking or transportation. The stopper is secured into a screw-threaded top joint via cap and Viton® O-ring for a positive, leaktight seal. Supplied complete with 1:5 PTFE plug, stopper, cap, and O-ring.
法律信息
Viton is a registered trademark of E. I. du Pont de Nemours and Company
历史批次信息供参考:
分析证书(COA)