主要文件
Z113077
Sigma-Aldrich
Aldrich® glass pH electrode
Standard pin connector
登录查看公司和协议定价
About This Item
UNSPSC代码:
41115603
NACRES:
NB.79
参数
−5-110 °C temperature
正在寻找类似产品? 访问 产品对比指南
一般描述
For applications where a separate reference electrode is required. Fast response over the full pH range with low sodium ion error. Useful for highly acidic or highly alkaline samples.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
相关产品
历史批次信息供参考:
分析证书(COA)