主要文件
SYNA464250
Synthware modified anti-splash adapter with fritted disc
250 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
别名:
Synthware A464250, Anti-splash adapter, splash head, bump trap
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121815
NACRES:
NB.51
接合处
bottom joint: ST/NS24/40
joint: ST/NS top24/40
包装
pkg of 1 ea
制造商/商品名称
Synthware A464250
容量
250 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
With extra coarse fritted disc sealed above lower inner joint. Frit reduces foam overflow from being drawn into the condenser.
法律信息
Synthware is a trademark of Kemtech America Inc.
历史批次信息供参考:
分析证书(COA)