主要文件
SLW1060/09D
Pyrex® Watch glasses
nominal diameter 60 mm
别名:
Pyrex, SciLabware 1060/09D
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121801
NACRES:
NB.43
包装
pkg of 10
制造商/商品名称
1060/09D
SciLabware
标称直径
60 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
Watch glasses
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
法律信息
PYREX is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)