主要文件
PL800GH-10
Plas-Labs Hypalon glove box gloves
for 1 piece glove set, size 10 (for 8 in. ports)
登录查看公司和协议定价
About This Item
UNSPSC代码:
46181504
NACRES:
NB.35
物料
Hypalon®
尺寸
10 (for 8 in. ports)
特点
reusable
包装
pack of 1 pair
制造商/商品名称
Plas-Labs 800GH-10
正在寻找类似产品? 访问 产品对比指南
一般描述
Plas-Labs white Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
法律信息
HaackerCalc is a trademark of ChemSW, Inc.
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
历史批次信息供参考:
分析证书(COA)