Merck
MT51302257
Mettler Toledo glass flow-through cell
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41115600
NACRES:
NB.61
材料
glass shaft
製造商/商標名
Mettler-Toledo 51302257
一般說明
Flow through cell for electrodes with 12 mm diameter