CLS410956
1 of 10
CLS410951
Corning® stir top plate assembly, 4x5
Sigma-Aldrich
CDS024515
4-[1,1′-Biphenyl]-4-yl-6-(4-chlorophenyl)-2-pyrimidinamine
CLS410933
Corning® stir top plate assembly, 5x7
C27115
对氯苄醇
Z742701
BenchMixer™ V2 Vortexer with reversible top
CLS410947
Corning® motor kit, PC-410/420, 230V
CLS410959
Corning® top plate assembly, PC-600, 120V
BMSH3770HE
Benchmark Digital 7 x 7 inch Hotplate
Z603929
Binder drying oven E28 series
Supelco
50652-U
Ascentis® Express 90 Å PCS-C18, 2.7 µm HPLC Column
Corning 410956
available only in South Korea
Corning®
Labware
Labware Partners