材料
colorless
glass
無菌
non-sterile
特點
without clips
製造商/商標名
BRAND 717810
長度 × 寬度 × 高度
24 mm × 26 mm × 0.4 mm
小室尺寸
0.1 mm , depth
孔
1
正在寻找类似产品? 访问 产品对比指南
一般說明
包括 2 个血球计数板盖玻片。尺寸为 24 mm Lx 26 mm Wx 0.4 mm H。它们也作为产品 BR723014 单独销售。
特點和優勢
- BLAUBRAND® 计数器是一种精密的测量仪器,用于测定每体积单位的悬浮粒子数。
- 与改进牛鲍型具有相同刻度,但计数器底部采用铑涂层。
- 刻度刻在铑涂层中,在正常显微镜下显得明亮。
- 通过改变对比度,可颠倒显微镜下的图像,进而根据需要使刻度看起来更亮或更暗。
包裝
包括 2 个血细胞计数板盖玻片。尺寸为 24 mm Lx 26 mm Wx 0.4 mm H。它们也作为产品 BR723014 单独销售。
法律資訊
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
其他客户在看
Biotechnology and bioengineering, 117(6), 1696-1709 (2020-02-27)
A barrier to cost-efficient biomanufacturing is the instability of engineered genetic elements, such as plasmids. Instability can also manifest at the whole-genome level, when fungal dikaryons revert to parental species due to nuclear segregation during cell division. Here, we show
Journal of cellular physiology, 231(3), 551-557 (2015-09-04)
Prolyl endopeptidase (PREP) is a serine protease which has been implicated in many biological processes, such as the maturation and degradation of peptide hormones and neuropeptides, learning and memory, cell proliferation and differentiation, and glucose metabolism. A small number of