主要文件
BAH942404004
Scienceware® Lab Companion perforated shelf
fits 11 L cabinet vacuum desiccators, pack of 1 ea
登录查看公司和协议定价
About This Item
UNSPSC代码:
41104510
NACRES:
NB.43
包装
pack of 1 ea
制造商/商品名称
Bel-Art H94240-4004
正在寻找类似产品? 访问 产品对比指南
一般描述
SP Bel-Art Perforated Shelf, available as a replacement of or addition to Lab Companion™ cabinet style vacuum desiccators. Perforated shelves permit efficient airflow throughout cabinet; all models allow for additional shelves to be added.
法律信息
Lab Companion is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)