主要文件
BAH188620000
Scienceware® tiered test tube rack
Holds 16 x 10-14 mm tubes
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122804
NACRES:
NB.35
材料
high-density polypropylene
特點
Holds 16 x 10-14 mm tubes
包裝
pack of 1 ea
製造商/商標名
Bel-Art H18862-0000
長度 × 寬度 × 高度
23.9 cm × 6.4 cm × 6.6 cm
尺寸
330x330x44 mm
一般說明
Compact, test tube rack takes up very little space and keeps up to 16 tubes upright and organized. Tiered for easier viewing.
  • 16 place rack holds tubes 10-14mm
  • Double tiered rack with solid bottom platform, keeps tubes stable
法律資訊
Scienceware is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)