主要文件
BAF169600000
SP Bel-Art® Truncated Style Acid/Solvent Bottle Carrier
to hold, one 4 Liter Bottle (1 gallon)
别名:
solvent bottle carrier
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122800
NACRES:
NB.43
特点
to hold one 4 Liter Bottle (1 gallon)
包装
pack of 1 ea
制造商/商品名称
Bel-Art F16960-0000
高度 × 直径
13.375 in. (350 mm) × 8 in. (200 mm)
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles.
特点和优势
  • Cone style lid has a 16mm (5/8”) opening in the top for insertion of tubing; simplifies the transfer of liquids without pouring
  • Truncated style lid permits pouring without the removal of the lid
  • Designed to hold standard US reagent bottles
法律信息
Bel-Art is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)