TLC Explorer赋能数字化分析
数字化TLC分析
薄层色谱(TLC)是一项广泛使用的化学分析技术,不过,色谱板的传统分析方法可能会比较麻烦。TLC Explorer系统提供一种定量效率和准确性更高的数字化解决方案,实现更加一致的结果和轻松的数据分享。
主要特性和优势
准确的TLC成像:高分辨率数字成像技术,不到2分钟时间对TLC、HPTLC和PLC板细致成像,减少主观和不一致的解读。自动轨道识别,可同时分析单块或多块板。
可靠的定量分析:通过图像光密度法将斑点亮度转化为数值数据,轻松定量。这种方法对于浓度测量和实验结果比较都更便利。
顺畅的流程衔接:无缝连接电脑、平板和智能手机,数据分享和存档高效。
用户友好设计,普通实验室中操作方便
- 3种色谱图LED照明光源:白光、UV 366 nm和UV 254 nm,从上方直接射光模式照亮色谱板,轻松鉴定物质
- 即插即用设置,搭配直观色谱软件
- 可通过观察窗进行快速的肉眼检查,免去紫外暴露风险,提高安全性
- 便携紧凑,方便离网运行
相关资源
- Brochure: TLC Explorer Documentation System
Your Thin-Layer Chromatography (TLC) experience more convenient whether in analytical routine, method development, in-process control, or research.
- User Manual: TLC Explorer Documentation System
The TLC Explorer is designed to document and evaluate thin-layer chromatographic plates.
- Application Note: Reaction Monitoring of the Esterification Progress of Benzoic Acid using Thin-Layer Chromatography (TLC)
Safe and secure UV handling using TLC Explorer Documentation System
- Application Note: Identification of Dexamethasone and its Derivatives by Thin-Layer Chromatography (TLC)
According to European Pharmacopoeia by using TLC Explorer Documentation System
设置安全保护,防止使用时紫外暴露。色谱板(单块或多块)牢牢固定在抽屉上(图1),仅当抽屉关闭后才会开始分析,确保无紫外暴露。另采用更环保的LED光源取代传统的汞灯。
Figure 1.Figure 1. TLC plates are positioned on a removable Baseplate (P1). The frame (P2) can be removed for easier handling. An alignment pin underneath the plate area (P3) helps the user to orient and align the Baseplate correctly on the Drawer (P4).
数字文档系统为整个TLC检测流程带来灵活性和便利性。可经由WiFi或二维码扫描轻松连接系统,便于在多种设备上运行(图2)。
Figure 2.Seamless operation of the TLC Explorer on a phone, laptop, and a tablet.
色谱板分析交由直观软件自动完成,无需繁琐耗时的人工数据处理和操作。自动识别轨道,可以不到2分钟完成轨道测量。软件还可同时检测多块板,覆盖面积高达20 x 20 cm(图3)。
Figure 3.Multiple TLC plates for analysis in the TLC Explorer.
即使在测量前色谱板未对准,该软件仍可自动旋转图像来对准。必要时还可使用背景降噪滤波器。该软件还可自动预选起始线和溶剂前沿,但也可选择手动添加和删除轨迹、溶剂和起始线。您可轻松添加注释进一步阐释,例如方法细节或命名规则。此外,该软件还可方便进行TLC和HPTLC色谱板对比和定量检查。
采用人体工学设计(图6a),可通过观察窗快速检查色谱板(图5)。观察窗通过挡板滑块(图4,F8)来控制开口,减少了用户通过窗口进行观察时入射杂散光的干扰。通过上下滑动滑块,便可查看底板的不同部分。
Figure 4.TLC Explorer, Documentation system. F1: Power Button; F2, F3, F4: Illumination Buttons and Indicators for manual use; F5, F6: Drawer and “Drawer open” Indicator; F7, F8: Inspection Window and Shuttler Slider for visual plate analysis.
Figure 5.Open inspection window for safe visual check. Image shows an illuminated plate under UV 254 nm excitation.
该设备配有三种激发光源,只需轻按相应波长的触控感应按钮,即可轻松开启光源(图4,F2–4)。但一次只能触发一种光源。
Figure 6.The front of the device has an angle of 12° (a) allowing a more ergonomic operation while sitting (b) and standing.